Learn More
2,2'-((2-Aminoethyl)azanediyl)diethanol, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 3197-06-6 |
| Molecular Formula | C6H16N2O2 |
| MDL Number | MFCD00014822 |
| Quantity | 1 g |
| SMILES | NCCN(CCO)CCO |
| Molecular Weight (g/mol) | 148.20 |
| Percent Purity | 95% |
| Chemical Name or Material | 2,2'-((2-Aminoethyl)azanediyl)diethanol |