missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. p-7029697
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15417665

Brand: Thermo Scientific Alfa Aesar H27190.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073354-41-2
Molecular Formula C15H23BN2O2
Molecular Weight (g/mol) 274.17
MDL Number MFCD06798280
InChI Key RKTXCUGUPDEZHR-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-yl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-pyrrolidin-1-yl pyridine-3-boronic acid pinacol ester, 2-1-pyrrolidinyl pyridine-3-boronic acid pinacol ester, 2-pyrrolidin-1-ylpyridine-3-boronic acid, pinacol ester, 2-pyrrolidinopyridine-3-boronic acid, pinacol ester, 2-1-pyrrolidino pyridine-3-boronic acid pinacol ester, 2-pyrrolidin-1-yl-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-1-pyrrolidinyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 44755171
IUPAC Name 2-pyrrolidin-1-yl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=C(N=CC=C1)N1CCCC1

Specifications

Melting Point 76°C to 78°C
Quantity 250 mg
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 274.17
Percent Purity 95%
Chemical Name or Material 2-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.