missing translation for 'onlineSavingsMsg'
Learn More

2-[(1-Hydroxy-2-oxo-2-phenylethyl)sulfanyl]acetic Acid, TRC

Product Code. 30406815
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406815 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406815 Supplier Toronto Research Chemicals Supplier No. TRCH95606550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(1-Hydroxy-2-oxo-2-phenylethyl)sulfanyl]acetic Acid
CAS 37510-29-5
Molecular Formula C10 H10 O4 S
Quantity 50 mg
Synonym 2-[(1-Hydroxy-2-oxo-2-phenylethyl)thio]acetic Acid
InChI Formula InChI=1S/C10H10O4S/c11-8(12)6-15-10(14)9(13)7-4-2-1-3-5-7/h1-5,10,14H,6H2,(H,11,12)
SMILES OC(SCC(=O)O)C(=O)c1ccccc1
IUPAC Name 2-(1-hydroxy-2-oxo-2-phenyl-ethyl)sulfanylacetic acid
Molecular Weight (g/mol) 226.25
Formula Weight 226.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.