Learn More
2-(1-Adamantyl)-4-bromoanisole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(1-Adamantyl)-4-bromoanisole |
| CAS | 104224-63-7 |
| Molecular Formula | C17H21BrO |
| Quantity | 250 mg |
| Synonym | 1-(5-Bromo-2-methoxyphenyl)-tricyclo[3.3.1.13,7]decane |
| InChI Formula | InChI=1S/C17H21BrO/c1-19-16-3-2-14(18)7-15(16)17-8-11-4-12(9-17)6-13(5-11)10-17/h2-3,7,11-13H,4-6,8-10H2,1H3/t11-,12+,13-,17- |
| SMILES | COc1ccc(Br)cc1[C@@]23C[C@@H]4C[C@@H](C[C@@H](C4)C2)C3 |
| IUPAC Name | 1-(5-bromo-2-methoxyphenyl)adamantane |
| Molecular Weight (g/mol) | 321.25 |
| Formula Weight | 320.0776 |