Learn More
(-)-(1S,4R)-Camphanoyl Chloride, TRC
Product Code. 30329386
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329386 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30329386 Supplier Toronto Research Chemicals Supplier No. TRCC1650001G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (-)-(1S,4R)-Camphanoyl Chloride
CAS 39637-74-6
Molecular Formula C10H13ClO3
Quantity 1 g
InChI Formula InChI=1S/C10H13ClO3/c1-8(2)9(3)4-5-10(8,6(11)12)14-7(9)13/h4-5H2,1-3H3/t9-,10+/m0/s1
SMILES CC1(C)[C@@]2(C)CC[C@@]1(OC2=O)C(=O)Cl
IUPAC Name (1R,4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride
Molecular Weight (g/mol) 216.66
Formula Weight 216.055