Learn More
(1R,4R)-1,8-Bis(diphenylacetoxy)-2-menthene, TRC
Specifications
Specifications
| Chemical Name or Material | (1R,4R)-1,8-Bis(diphenylacetoxy)-2-menthene |
| CAS | 477528-49-7 |
| Molecular Formula | C38H38O4 |
| Quantity | 50 mg |
| Synonym | alpha-Phenyl-benzeneacetic Acid 1-[(1R,4R)-4-[(Diphenylacetyl)oxy]-4-methyl-2-cyclohexen-1-yl]-1-methylethyl Ester |
| InChI Formula | InChI=1S/C38H38O4/c1-37(2,41-35(39)33(28-16-8-4-9-17-28)29-18-10-5-11-19-29)32-24-26-38(3,27-25-32)42-36(40)34(30-20-12-6-13-21-30)31-22-14-7-15-23-31/h4-24,26,32-34H,25,27H2,1-3H3/t32-,38-/m0/s1 |
| SMILES | CC(C)(OC(=O)C(c1ccccc1)c2ccccc2)[C@@H]3CC[C@@](C)(OC(=O)C(c4ccccc4)c5ccccc5)C=C3 |
| IUPAC Name | [(1R,4R)-4-[2-(2,2-diphenylacetyl)oxypropan-2-yl]-1-methylcyclohex-2-en-1-yl] 2,2-diphenylacetate |
| Molecular Weight (g/mol) | 558.71 |
| Formula Weight | 558.277 |