Learn More
(1R)-3-endo-Aminoborneol, TRC
Specifications
Specifications
| Chemical Name or Material | (1R)-3-endo-Aminoborneol |
| CAS | 29900-93-4 |
| Molecular Formula | C10H19NO |
| Quantity | 5 mg |
| Synonym | endo-3-Amino-d-borneol, (+)-endo-2-Hydroxy-endo-3-aminobornane, (1R)-3-endo-Amino-2-endo-bornanol, [1R-(endo,endo)]-3-Amino-1,7,7-trimethylbiicyclo[2.2.1]heptan-2-ol, (1R,2R,3S,4S)-3-Amino-2-bornanol |
| InChI Formula | InChI=1S/C10H19NO/c1-9(2)6-4-5-10(9,3)8(12)7(6)11/h6-8,12H,4-5,11H2,1-3H3/t6?,7-,8+,10?/m0/s1 |
| SMILES | CC1(C)C2CCC1(C)[C@H](O)[C@H]2N |
| IUPAC Name | (2S,3S)-2-amino-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol |
| Molecular Weight (g/mol) | 169.26 |
| Formula Weight | 169.1467 |