Learn More
(1R)-3-endo-Aminoborneol, TRC
Product Code. 30326271
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326271 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326271 Supplier Toronto Research Chemicals Supplier No. TRCA62257025MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (1R)-3-endo-Aminoborneol
CAS 29900-93-4
Molecular Formula C10H19NO
Quantity 25 mg
Synonym endo-3-Amino-d-borneol, (+)-endo-2-Hydroxy-endo-3-aminobornane, (1R)-3-endo-Amino-2-endo-bornanol, [1R-(endo,endo)]-3-Amino-1,7,7-trimethylbiicyclo[2.2.1]heptan-2-ol, (1R,2R,3S,4S)-3-Amino-2-bornanol
InChI Formula InChI=1S/C10H19NO/c1-9(2)6-4-5-10(9,3)8(12)7(6)11/h6-8,12H,4-5,11H2,1-3H3/t6?,7-,8+,10?/m0/s1
SMILES CC1(C)C2CCC1(C)[C@H](O)[C@H]2N
IUPAC Name (2S,3S)-2-amino-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
Molecular Weight (g/mol) 169.26
Formula Weight 169.1467