Learn More
(1R,2R,1’R,2’S)-Atracurium Besylate, >90%, TRC
Specifications
Specifications
| Chemical Name or Material | R-cis-R'-trans-Atracurium Besylate |
| CAS | 96946-44-0 |
| Molecular Formula | C53 H72 N2 O12 . 2 C6 H5 O3 S |
| Quantity | 5 mg |
| Synonym | (1R,2R,1'R,2'S)-Atracurium Besylate, Isoquinolinium, 2,2'-[1,5-pentanediylbis[oxy(3-oxo-3,1-propanediyl)]]bis[1-[(3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-, (1R,1'R,2R,2'S)-, benzenesulfonate (1:2), Isoquinolinium, 2,2'-[1,5-pentanediylbis[oxy(3-oxo-3,1-propanedi |
| InChI Formula | InChI=1S/C53H72N2O12.2C6H6O3S/c1-54(22-18-38-32-48(62-7)50(64-9)34-40(38)42(54)28-36-14-16-44(58-3)46(30-36)60-5)24-20-52(56)66-26-12-11-13-27-67-53(57)21-25-55(2)23-19-39-33-49(63-8)51(65-10)35-41(39)43(55)29-37-15-17-45(59-4)47(31-37)61-6;2*7-10(8,9)6-4 |
| SMILES | COc1ccc(C[C@@H]2c3cc(OC)c(OC)cc3CC[N@+]2(C)CCC(=O)OCCCCCOC(=O)CC[N@+]4(C)CCc5cc(OC)c(OC)cc5[C@H]4Cc6ccc(OC)c(OC)c6)cc1OC.[O-]S(=O)(=O)c7ccccc7.[O-]S(=O)(=O)c8ccccc8 |
| IUPAC Name | benzenesulfonate;5-[3-[(1R,2R)-1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinolin-2-ium-2-yl]propanoyloxy]pentyl 3-[(1R,2S)-1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinolin-2-ium-2-yl]propano |
| Molecular Weight (g/mol) | 1243.479 |
| Formula Weight | 1242.5 |