missing translation for 'onlineSavingsMsg'
Learn More

[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]monophenylester, TRC

Product Code. 30314819
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
missing translation for 'unitSize'
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314819 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30314819 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCA628898250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [((1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy)methyl]monophenylester
CAS 379270-35-6
Molecular Formula C15H18N5O4P
Quantity 250 mg
Synonym Phenyl Hydrogen [(R)-1-(6-Amino-9H-purin-9-yl)propan-2-yloxy]methylphosphonate
InChI Formula InChI=1S/C15H18N5O4P/c1-11(7-20-9-19-13-14(16)17-8-18-15(13)20)23-10-25(21,22)24-12-5-3-2-4-6-12/h2-6,8-9,11H,7,10H2,1H3,(H,21,22)(H2,16,17,18)/t11-/m1/s1
SMILES NC1=C2C(N(C[C@@H](C)OCP(OC3=CC=CC=C3)(O)=O)C=N2)=NC=N1
IUPAC Name [(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-phenoxyphosphinic acid
Molecular Weight (g/mol) 363.31
Formula Weight 363.1096
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.