Learn More
(1R)-(-)-(10-Camphorsulfonyl)oxaziridine, TRC
Specifications
Specifications
| Chemical Name or Material | (1R)-(-)-(10-Camphorsulfonyl)oxaziridine |
| CAS | 104372-31-8 |
| Molecular Formula | C10 H15 N O3 S |
| Quantity | 25 g |
| InChI Formula | InChI=1S/C10H15NO3S/c1-8(2)7-3-4-9(8)6-15(12,13)11-10(9,5-7)14-11/h7H,3-6H2,1-2H3/t7-,9-,10?,11?/m1/s1 |
| SMILES | CC1(C)[C@@H]2CC[C@@]13CS(=O)(=O)N4OC34C2 |
| IUPAC Name | 11,11-dimethyl-5-oxa-3λ6-thia-4-azatetracyclo[6.2.1.01,6.04,6]undecane 3,3-dioxide |
| Molecular Weight (g/mol) | 229.3 |
| Formula Weight | 229.0773 |