missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R)-1-((4R,4aR,8aS)-2,6-Bis(3,4-dimethylphenyl)tetrahydro-[1,3]dioxino[5,4-d][1,3]dioxin-4-yl)ethane-1,2-diol, TRC
Specifications
Specifications
| Chemical Name or Material | D-Sorbit-bis(3,4-dimethylbenzylidene) |
| CAS | 135861-56-2 |
| Molecular Formula | C24 H30 O6 |
| Quantity | 1 g |
| Synonym | 1,3:2,4-Bis-O-[(3,4-dimethylphenyl)methylene]-D-glucitol, [1,3]Dioxino[5,4-d]-1,3-dioxin, D-glucitol deriv., 1,3:2,4-Bis(3,4-dimethylbenzylidene)sorbitol, 1,3:2,4-Bis(3',4'-dimethylbenzylidene)sorbitol, 1,3:2,4-Bis-O-(3,4-dimethylbenzylidene)-D-sorbitol, 1,3:2,4-Di(3,4-dimethylbenzylidene)-D-sorbitol, 1,3:2,4-Di(3,4-dimethylbenzylidene)sorbitol, 1,3:2,4-Di-O-3,4-dimethylbenzylidene-D-sorbitol, Bis(3,4-dimethylbenzylidene) sorbitol, Chiclear 390, DMDBS, Geniset DXR, Jadewin 3988, Millad 3988, Millad 3988I, Millad 3998, Millad 8C41-10, Nucleating Agent 3988, ST-NC 38, TH 3988, ZC 3 |
| InChI Formula | InChI=1S/C24H30O6/c1-13-5-7-17(9-15(13)3)23-27-12-20-22(30-23)21(19(26)11-25)29-24(28-20)18-8-6-14(2)16(4)10-18/h5-10,19-26H,11-12H2,1-4H3/t19-,20+,21-,22-,23?,24?/m1/s1 |
| SMILES | Cc1ccc(cc1C)C2OC[C@@H]3OC(O[C@H]([C@H](O)CO)[C@@H]3O2)c4ccc(C)c(C)c4 |
| IUPAC Name | (1R)-1-[(4R,4aR,8aS)-2,6-bis(3,4-dimethylphenyl)-4,4a,8,8a-tetrahydro-[1,3]dioxino[5,4-d][1,3]dioxin-4-yl]ethane-1,2-diol |
| Molecular Weight (g/mol) | 414.49 |
| Formula Weight | 414.2042 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?