Learn More
(1R)-1-(3,4-Dimethoxyphenyl)-1,2-ethanediol, TRC
Product Code. 30382751
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382751 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30382751 Supplier Toronto Research Chemicals Supplier No. TRCD461250250MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (1R)-1-(3,4-Dimethoxyphenyl)-1,2-ethanediol
CAS 326491-79-6
Molecular Formula C10H14O4
Quantity 250 mg
InChI Formula InChI=1S/C10H14O4/c1-13-9-4-3-7(8(12)6-11)5-10(9)14-2/h3-5,8,11-12H,6H2,1-2H3/t8-/m0/s1
SMILES COc1ccc(cc1OC)[C@@H](O)CO
Molecular Weight (g/mol) 198.22
Formula Weight 198.0892