missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, 11⁄4Cr 1⁄2Mo, A182 Grade F11, K11572, 100g, ARMI∣MBH
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Ea.
missing translation for 'unitSize'
Each
Description
Alloy Steel, 11⁄4Cr 1⁄2Mo, A182 Grade F11, K11572, 100g
Specifications
Specifications
| Chemical Name or Material | Nitrogen/Tungsten/Niobium/Zirconium/Lead/Boron/Titanium/Antimony/Vanadium/Arsenic/Cobalt/Phosphorus/Tin/Sulfur/Aluminium/Nickel/Carbon/Copper/Manganese/Molybdenum/Silicon/Chromium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Nitrogen -80 ppm/Tungsten -30 ppm/Niobium -20 ppm/Zirconium -20 ppm/Lead -10 ppm/Boron 0.0004 %/Titanium 0.0015 %/Antimony 0.0017 %/Vanadium 0.0038 %/Arsenic 0.0048 %/Cobalt 0.0062 %/Phosphorus 0.0067 %/Tin 0.0084 %/Sulfur 0.0141 %/Aluminium 0.018 %/Nickel 0.069 %/Carbon 0.114 %/Copper 0.123 %/Manganese 0.53 %/Molybdenum 0.614 %/Silicon 0.679 %/Chromium 1.349 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction