missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Manganese Bronze, CDA 863, C86300, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Manganese Bronze, CDA 863, C86300, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Zirconium/Magnesium/Cadmium/Cobalt/Antimony/Bismuth/Silver/Carbon/Arsenic/Chromium/Phosphorus/Silicon/Lead/Tin/Nickel/Iron/Manganese/Aluminium/Zinc/Copper/Beryllium/Selenium/Boron/Tellurium/Nitrogen/Oxygen/Niobium |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Zirconium 0.0004 %/Magnesium 0.0006 %/Cadmium 0.0009 %/Cobalt 0.001 %/Antimony 0.003 %/Bismuth 0.004 %/Silver 0.004 %/Carbon 0.005 %/Arsenic 0.007 %/Chromium 0.008 %/Phosphorus 0.02 %/Silicon 0.091 %/Lead 0.133 %/Tin 0.147 %/Nickel 0.276 %/Iron 2.98 %/Manganese 2.99 %/Aluminium 5.79 %/Zinc 22.86 %/Copper 64.5 %/Beryllium (<0.005) %/Selenium (<0.005) %/Boron (<0.1) %/Tellurium (0.0001) %/Nitrogen (0.00016) %/Oxygen (0.0006) %/Niobium (0.001) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction