missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Chromium Copper, CDA 182, C18200, 100g, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Chromium Copper, CDA 182, C18200, 100g
Specifications
Specifications
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction