missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, A36, K02600, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, A36, K02600, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Vanadium/Titanium/Niobium/Aluminium/Tungsten/Nitrogen/Cobalt/Phosphorus/Molybdenum/Tin/Nickel/Chromium/Carbon/Copper/Silicon/Manganese/Lead/Tantalum/Boron/Calcium/Zinc/Antimony/Arsenic/Oxygen |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Vanadium 0.001 %/Titanium 0.0011 %/Niobium 0.0012 %/Aluminium 0.003 %/Tungsten 0.003 %/Nitrogen 0.008 %/Cobalt 0.009 %/Phosphorus 0.012 %/Molybdenum 0.0131 %/Tin 0.0147 %/Nickel 0.076 %/Chromium 0.093 %/Carbon 0.158 %/Copper 0.207 %/Silicon 0.226 %/Manganese 0.725 %/Lead (<0.001) %/Tantalum (<0.005) %/Boron (0.0004) %/Calcium (0.0015) %/Zinc (0.002) %/Antimony (0.0032) %/Arsenic (0.006) %/Oxygen (0.01) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction