Learn More
Veraguensin, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Veraguensin |
| CAS | 19950-55-1 |
| Molecular Formula | C22H28O5 |
| Quantity | 1 mg |
| Synonym | Veraguensin, (2R,3S,4S,5S)-2,5-Bis(3,4-dimethoxyphenyl)tetrahydro-3,4-dimethylfuran, (+)-Veraguensin, Veraguensine |
| InChI Formula | InChI=1S/C22H28O5/c1-13-14(2)22(16-8-10-18(24-4)20(12-16)26-6)27-21(13)15-7-9-17(23-3)19(11-15)25-5/h7-14,21-22H,1-6H3/t13-,14-,21-,22+/m0/s1 |
| SMILES | O=C(NC(C(N)=O)C(C)(C)C)C1=NN(CC2CCCCC2)C3=C1C=CC=C3 |
| Molecular Weight (g/mol) | 372.45 |