missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiobencarb-d10 (diethyl-d10), TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Thiobencarb diethyl-d10 |
| CAS | 1219804-12-2 |
| Molecular Formula | C12 D10 H6 Cl N O S |
| Quantity | 1 mg |
| Synonym | Carbamothioic acid','N,N-di(ethyl-1,1,2,2,2-d5)-','S-[(4-chlorophenyl)methyl] ester (ACI), S-[(4-Chlorophenyl)methyl] N,N-di(ethyl-1,1,2,2,2-d5)carbamothioate (ACI), Thiobencarb-d10, N,N-Di(ethyl-1,1,2,2,2-d5)-S-[(4-Chlorophenyl)methyl] Ester Carbamothioic Acid |
| InChI Formula | InChI=1S/C12H16ClNOS/c1-3-14(4-2)12(15)16-9-10-5-7-11(13)8-6-10/h5-8H,3-4,9H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])N(C(=O)SCc1ccc(Cl)cc1)C([2H])([2H])C([2H])([2H])[2H] |
| IUPAC Name | S-[(4-chlorophenyl)methyl] N,N-bis(1,1,2,2,2-pentadeuterioethyl)carbamothioate |
| Molecular Weight (g/mol) | 267.84 |
| Formula Weight | 267.1269 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction