missing translation for 'onlineSavingsMsg'
Learn More

Pyochelin I and Pyochelin II, TRC

Product Code. 30009078
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009078 1 mg 1mg
30008898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009078 Supplier Toronto Research Chemicals Supplier No. TRCP8403651MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79236-62-7
Molecular Formula C14H16N2O3S2
Molecular Weight (g/mol) 324.42
Synonym (4R)-2-((R)-2-(2-hydroxyphenyl)-4,5-dihydrothiazol-4-yl)-3-methylthiazolidine-4-carboxylic Acid, mix of Pyochelin I and Pyochelin II
SMILES >>OC1=CC=CC=C1C2=N[C@]([C@@]3([H])N(C)[C@H](C(O)=O)CS3)([H])CS2

Specifications

Quantity 1 mg
Chemical Name or Material Pyochelin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.