missing translation for 'onlineSavingsMsg'
Learn More

Cefixime-13C,15N2, TRC

Product Code. 17941375
Change view
Click to view available options
Quantity:
0.25 mg
1 mg
2.5 mg
Unit Size:
0.25mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17941375 1 mg 1mg
30132673 2.5 mg 2.5mg
30135941 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17941375 Supplier Toronto Research Chemicals Supplier No. TRCC2428021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C1513CH15N315N2O7S2
Molecular Weight (g/mol) 456.43
Synonym (6R,7R)-7-[[(2Z)-2-(2-Amino-4-thiazolyl-13C,15N2)-2-[(carboxymethoxy)imino]acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, FK-027-13C,15N2, FR- 17027-13C,15N2, CL-284635-13C,15N2;Cefixoral-13C,15N2, Cefspan-13C,15N2, Cephoral-13C,15N2, Unixime-13C,15N2;
IUPAC Name (6R,7R)-7-[[(2E)-2-(2-(15N)azanyl-(213C,315N)1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES C=CC1=C(C(O)=O)N2[C@]([C@]([H])(NC(/C(C3=CS[13C]([15NH2])=[15N]3)=N\OCC(O)=O)=O)C2=O)([H])SC1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C16H15N5O7S2/c1-2-6-4-29-14-10(13(25)21(14)11(6)15(26)27)19-12(24)9(20-28-3-8(22)23)7-5-30-16(17)18-7/h2,5,10,14H,1,3-4H2,(H2,17,18)(H,19,24)(H,22,23)(H,26,27)/b20-9+/t10-,14-/m1/s1/i16+1,17+1,18+1
Percent Purity >95%
Chemical Name or Material Cefixime-13C,15N2
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.