Learn More
Bisoprolol-d7 Hemifumarate, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Bisoprolol-d7 Hemifumarate |
| CAS | 1310012-16-8 |
| Molecular Formula | 2[C18 D7 H24 N O4] . C4 H4 O4 |
| Quantity | 1 mg |
| Synonym | 1-(1,1,1,2,3,3,3-Heptadeuteriopropan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol hemifumarate, Bisoprolol-d7 hemifumarate, Bisoprolol-D7 (N-isopropyl-D7) hemifumarate, 1-[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]-3-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]-2-propanol (2E)-2-butenedioate (2:1) |
| InChI Formula | InChI=1S/2C18H31NO4.C4H4O4/c2*1-14(2)19-11-17(20)13-23-18-7-5-16(6-8-18)12-21-9-10-22-15(3)4;5-3(6)1-2-4(7)8/h2*5-8,14-15,17,19-20H,9-13H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+/i2*1D3,2D3,14D; |
| SMILES | [2H]C([2H])([2H])C([2H])(NCC(O)COc1ccc(COCCOC(C)C)cc1)C([2H])([2H])[2H].[2H]C([2H])([2H])C([2H])(NCC(O)COc2ccc(COCCOC(C)C)cc2)C([2H])([2H])[2H].OC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-but-2-enedioic acid;1-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol |
| Molecular Weight (g/mol) | 781.0445 |
| Formula Weight | 780.5495 |