Learn More
Thermo Scientific Alfa Aesar 1LT THF, anh., 99.8+%, stab. with 0.025% BHT, pack
Brand:  Thermo Scientific Alfa Aesar 047382.K2
1LT THF, anh., 99.8+%, stab. with 0.025% BHT, pack. under Ar in resealable AcroSeal bttls
Product Code. 16358227
  • 94.50€ / 1 L