missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,2H,2H-Perfluorododecyl Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | 1H,1H,2H,2H-Perfluorododecane Acetate |
| CAS | 37858-05-2 |
| Purity Grade Notes | GC |
| Molecular Formula | C14 H7 F21 O2 |
| Quantity | 50 mg |
| Synonym | 1H,1H,2H,2H-Perfluorododecyl Acetate, 1-Dodecanol','3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-heneicosafluoro-','1-acetate (ACI), 1-Dodecanol','3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-heneicosafluoro-','acetate (9CI) |
| InChI Formula | InChI=1S/C14H7F21O2/c1-4(36)37-3-2-5(15,16)6(17,18)7(19,20)8(21,22)9(23,24)10(25,26)11(27,28)12(29,30)13(31,32)14(33,34)35/h2-3H2,1H3 |
| SMILES | CC(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC Name | 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecyl acetate |
| Molecular Weight (g/mol) | 606.171 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?