Learn More
Valganciclovir Hydrochloride, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Valganciclovir Hydrochloride |
| CAS | 175865-59-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 H22 N6 O5 . Cl H |
| Quantity | 1 g |
| Synonym | L-Valine','2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-hydroxypropyl ester','hydrochloride (1:1), L-Valine','2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-hydroxypropyl ester','monohydrochloride (9CI), RS 079070-194, Ro 107-9070/194, Valcyte, Valganciclovir hydrochloride |
| InChI Formula | InChI=1S/C14H22N6O5.ClH/c1-7(2)9(15)13(23)24-4-8(3-21)25-6-20-5-17-10-11(20)18-14(16)19-12(10)22;/h5,7-9,21H,3-4,6,15H2,1-2H3,(H3,16,18,19,22);1H/t8?,9-;/m0./s1 |
| SMILES | Cl.CC(C)[C@H](N)C(=O)OCC(CO)OCn1cnc2C(=O)N=C(N)Nc12 |
| IUPAC Name | [2-[(2-amino-6-oxo-3H-purin-9-yl)methoxy]-3-hydroxypropyl] (2S)-2-amino-3-methylbutanoate;hydrochloride |
| Molecular Weight (g/mol) | 390.82 |
| Show More |