missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Styrene-beta,beta-d2, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Styrene beta,beta-d2 |
| CAS | 934-85-0 |
| Molecular Formula | C8 2H2 H6 |
| Quantity | 1 g |
| Synonym | Benzene, ethenyl-2,2-d2-, Styrene-β,β-d2 (6 CI,7 CI,8 CI), Ethenyl-2,2-d2benzene, Styrene-β-d2, β,β-Dideuteriostyrene, Styrene-β,β-D2 |
| InChI Formula | InChI=1 S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7 H,1H2/i1D2 |
| SMILES | [2 H]C(=Cc1ccccc1)[2 H] |
| IUPAC Name | 2,2-dideuterioethenylbenzene |
| Molecular Weight (g/mol) | 106.16 |
| Formula Weight | 106.0752 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction