missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propio-2,2-d2-phenone, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Propiophenone-D2 (ethyl-1,1-D2) |
| CAS | 129848-87-9 |
| Molecular Formula | C9 D2 H8 O |
| Quantity | 1 g |
| Synonym | 1-Propanone-2,2-d2, 1-phenyl- (9 CI, ACI), 1-Phenyl-1-propanone-2,2-d2 (ACI), Propiophenone-d2, Propiophenone-2,2-d2, Propiophenone-D2 (ethyl-1,1-D2) |
| InChI Formula | InChI=1 S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i2D2 |
| SMILES | [2 H]C([2 H])(C)C(=O)c1ccccc1 |
| IUPAC Name | 2,2-dideuterio-1-phenylpropan-1-one |
| Molecular Weight (g/mol) | 136.187 |
| Formula Weight | 136.086 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction