missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecanoic-d35 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Stearic-D35 Acid |
| CAS | 17660-51-4 |
| Molecular Formula | C18 D35 H O2 |
| Quantity | 1 g |
| Synonym | Octadecanoic-D35 acid, Octadecanoic-d35 acid (9 CI, ACI), Stearic-d35 acid (8 CI), Stearic acid-2H35, Octadecanoic acid D35 |
| InChI Formula | InChI=1 S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,18-pentatriacontadeuteriooctadecanoic acid |
| Molecular Weight (g/mol) | 319.69 |
| Formula Weight | 319.4912 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction