Learn More
n-Hexyl-2,2,3,3,4,4,5,5,6,6,6-d11 Alcohol, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Hexanol-2,2,3,3,4,4,5,5,6,6,6-D11 |
| CAS | 2159-18-4 |
| Molecular Formula | C6 D11 H3 O |
| Quantity | 1 g |
| Synonym | 1-Hexanol-d11, 1-Hexan-2,2,3,3,4,4,5,5,6,6,6-d11-ol (ACI), Hexyl-2,2,3,3,4,4,5,5,6,6,6-d11 alcohol (7 CI, 8 CI), 1-Hexanol-d11, 1-Hexanol-2,2,3,3,4,4,5,5,6,6,6-D11, n-Hexyl-2,2,3,3,4,4,5,5,6,6,6-d11 alcohol, 1-Hexanol-2,2,3,3,4,4,5,5,6,6,6-d11 |
| InChI Formula | InChI=1 S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CO |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,6-undecadeuteriohexan-1-ol |
| Molecular Weight (g/mol) | 113.243 |
| Formula Weight | 113.174 g/mol |
| Show More |