Learn More
n-Butylbenzene-2,3,4,5,6-d5, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Butylbenzene-D5 (phenyl-D5) |
| CAS | 20329-91-3 |
| Molecular Formula | C10 D5 H9 |
| Quantity | 1 g |
| Synonym | Benzene-d5, butyl- (8 CI, 9 CI), 6-Butylbenzene-1,2,3,4,5-d5, Butylbenzene-d5, Butylbenzene-d5, Butylbenzene-D5 (phenyl-D5), n-Butylbenzene-2,3,4,5,6-d5 |
| InChI Formula | InChI=1 S/C10H14/c1-2-3-7-10-8-5-4-6-9-10/h4-6,8-9 H,2-3,7H2,1H3/i4D,5 D,6 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CCCC)c([2 H])c1[2 H] |
| IUPAC Name | 1-butyl-2,3,4,5,6-pentadeuteriobenzene |
| Molecular Weight (g/mol) | 139.249 |
| Formula Weight | 139.141 g/mol |
| Show More |