missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Bromoacetate-2,2-d2, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Methyl Bromoacetate-2,2-d2 |
| CAS | 163886-16-6 |
| Molecular Formula | C3 D2 H3 Br O2 |
| Quantity | 1 g |
| Synonym | Acetic-d2 acid, 2-bromo-, methyl ester (ACI), Acetic-d2 acid, bromo-, methyl ester (9 CI), Methyl 2-bromoacetate-d2 (ACI) |
| InChI Formula | InChI=1 S/C3H5BrO2/c1-6-3(5)2-4/h2H2,1H3/i2D2 |
| SMILES | [2 H]C([2 H])(Br)C(=O)OC |
| IUPAC Name | methyl 2-bromo-2,2-dideuterioacetate |
| Molecular Weight (g/mol) | 154.9869 |
| Formula Weight | 153.9598 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction