missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine-d3 (methyl-d3), CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | L-Leucine-d3 |
| CAS | 87828-86-2 |
| Molecular Formula | C6 2H3 H10 N O2 |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5 H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1/i1D3/t4?,5- |
| SMILES | [2 H]C([2 H])([2 H])C(C)C[C@H](N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-5,5,5-trideuterio-4-methylpentanoic acid |
| Molecular Weight (g/mol) | 134.19 |
| Formula Weight | 134.1135 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction