Learn More
Iodobenzene-3,5-d2, CDN
Product Code. 30322047
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322047
Brand: CDN CDND70031G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Iodobenzene-3,5-D2
CAS 51209-48-4
Molecular Formula C6 D2 H3 I
Quantity 1 g
Synonym Benzene-1,3-d2, 5-iodo- (9 CI), 1,3-Dideuterio-5-iodo-benzene, Iodobenzene-3,5-d2, Iodobenzene-3,5-D2
InChI Formula InChI=1 S/C6H5I/c7-6-4-2-1-3-5-6/h1-5 H/i2D,3 D
SMILES [2 H]c1cc([2 H])cc(I)c1
IUPAC Name 1,3-dideuterio-5-iodobenzene
Molecular Weight (g/mol) 206.0207
Formula Weight 205.9561 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity