missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroquinone-d6, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Hydroquinone (D6) |
| CAS | 71589-26-9 |
| Molecular Formula | C6 D6 O2 |
| Quantity | 1 g |
| Synonym | 1,4-Benzene-2,3,5,6-d4-diol-d2 (9 CI, ACI), 1,2,4,5-Tetradeuterio-3,6-dideuteriooxybenzene, 1,4-Hydroquinone-d6, Hydroquinone-d6, Perdeuteriohydroquinone, Hydroquinone-D6 |
| InChI Formula | InChI=1 S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8 H/i1D,2 D,3 D,4 D/hD2 |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c(O[2 H])c([2 H])c1[2 H] |
| IUPAC Name | 1,2,4,5-tetradeuterio-3,6-dideuteriooxybenzene |
| Molecular Weight (g/mol) | 116.1476 |
| Formula Weight | 116.0744 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction