missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine Sulfate-d6, CDN
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Hydrazine Sulfate-D6 |
| CAS | 24310-86-9 |
| Molecular Formula | D4 N2 . D2 O4 S |
| Quantity | 1 g |
| Synonym | Sulfuric acid-d2, compd. with hydrazine-d4 (1:1) (8 CI, 9 CI), Hydrazine-d4, sulfate-d2 (1:1) (8 CI, 9 CI), Hydrazine Sulfate-d6, Hydrazine Sulfate-D6 |
| InChI Formula | InChI=1 S/H4N2.H2O4S/c1-2;1-5(2,3)4/h1-2H2;(H2,1,2,3,4)/i/hD6 |
| SMILES | [2 H]OS(=O)(=O)O[2 H].[2 H]N([2 H])N([2 H])[2 H] |
| IUPAC Name | dideuterio sulfate;1,1,2,2-tetradeuteriohydrazine |
| Molecular Weight (g/mol) | 136.1606 |
| Formula Weight | 136.0425 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction