missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanoic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Hexanoic-2,2-d2 Acid |
| CAS | 55320-65-5 |
| Molecular Formula | C6 D2 H10 O2 |
| Quantity | 1 g |
| Synonym | Hexanoic-2,2-d2 acid (9 CI), 2,2-Dideuteriohexanoic acid, Hexanoic acid-d2, Hexanoic-α,α-d2 acid, Hexanoic-2,2-D2 acid, Caproic-α-d2 acid |
| InChI Formula | InChI=1 S/C6H12O2/c1-2-3-4-5-6(7)8/h2-5H2,1H3,(H,7,8)/i5D2 |
| SMILES | [2 H]C([2 H])(CCCC)C(=O)O |
| IUPAC Name | 2,2-dideuteriohexanoic acid |
| Molecular Weight (g/mol) | 118.171 |
| Formula Weight | 118.096 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction