missing translation for 'onlineSavingsMsg'
Learn More

Ferronil, TRC

Product Code. 30006525
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006525 1 g 1g
30134694 5 g 5g
30009012 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006525 Supplier Toronto Research Chemicals Supplier No. TRCF2574551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-71-3
Molecular Formula FeCO3
Molecular Weight (g/mol) 115.85
Synonym Ferrous Carbonate, Ferrous Monocarbonate, Iron Carbonate (FeCO3), Iron(2+) Carbonate, Iron(II) Carbonate;
IUPAC Name iron(2+);carbonate
SMILES [Fe+2].[O-]C(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/CH2O3.Fe/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2
Formula Weight 115.9197
Chemical Name or Material Ferronil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.