Learn More
Aniline-d7, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Aniline-d7 |
| CAS | 14545-23-4 |
| Molecular Formula | C6 2H7 N |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C6H7N/c7-6-4-2-1-3-5-6/h1-5 H,7H2/i1D,2 D,3 D,4 D,5 D/hD2 |
| SMILES | [2 H]N([2 H])c1c([2 H])c([2 H])c([2 H])c([2 H])c1[2 H] |
| IUPAC Name | N,N,2,3,4,5,6-heptadeuterioaniline |
| Molecular Weight (g/mol) | 100.17 |
| Formula Weight | 100.1018 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |