missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrotoluene-d7, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | 4-Nitrotoluene-d7 |
| CAS | 84344-19-4 |
| Molecular Formula | C7 2H7 N O2 |
| Quantity | 1 g |
| Synonym | Benzene-1,2,4,5-d4, 3-(methyl-d3)-6-nitro-, 3-(Methyl-d3)-6-nitrobenzene-1,2,4,5-d4, 4-Nitrotoluene-d7, p-Nitrotoluene-d7, 4-Nitrotoluene D7 |
| InChI Formula | InChI=1 S/C7H7NO2/c1-6-2-4-7(5-3-6)8(9)10/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1[N+](=O)[O-])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,4,5-tetradeuterio-3-nitro-6-(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 144.18 |
| Formula Weight | 144.0916 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur