Learn More
4-Methoxytoluene-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Methoxy-4-methylbenzene-2,3,5,6-D4 |
| CAS | 350818-57-4 |
| Molecular Formula | C8 D4 H6 O |
| Quantity | 1 g |
| Synonym | Benzene-1,2,4,5-d4, 3-methoxy-6-methyl- (9 CI), 4-Methoxytoluene-2,3,5,6-d4, p-Methylanisole-d4, 1-Methoxy-4-methylbenzene-2,3,5,6-d4, 1-Methoxy-4-methylbenzene-2,3,5,6-D4 |
| InChI Formula | InChI=1 S/C8H10O/c1-7-3-5-8(9-2)6-4-7/h3-6 H,1-2H3/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(OC)c([2 H])c([2 H])c1C |
| IUPAC Name | 1,2,4,5-tetradeuterio-3-methoxy-6-methylbenzene |
| Molecular Weight (g/mol) | 126.1891 |
| Formula Weight | 126.0983 g/mol |
| Show More |