Learn More
4-Methoxybenzaldehyde-alpha-d1, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 4-Methoxybenzaldehyde-D (formyl-D) |
| CAS | 19486-71-6 |
| Molecular Formula | C8 D H7 O2 |
| Quantity | 1 g |
| Synonym | Benzaldehyde-formyl-d, 4-methoxy- (9 CI, ACI), 4-Methoxybenzaldehyde-formyl-d (ACI), p-Anisaldehyde-formyl-d (8 CI), 4-Methoxy-[α-d]benzaldehyde, 4-Methoxybenzaldehyde-1-d, d1-Anisaldehyde, p-Anisaldehyde-1-d, p-Methoxybenzaldehyde-d1, p-Methoxybenzaldehyde-α-d, 4-Methoxybenzaldehyde-D (formyl-D) |
| InChI Formula | InChI=1 S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6 H,1H3/i6D |
| SMILES | [2 H]C(=O)c1ccc(OC)cc1 |
| IUPAC Name | deuterio-(4-methoxyphenyl)methanone |
| Molecular Weight (g/mol) | 137.1541 |
| Formula Weight | 137.0587 g/mol |
| Show More |