Learn More
3-Methylpyridine-d7, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 3-Picoline-d7 |
| CAS | 202529-13-3 |
| Molecular Formula | C6 D7 N |
| Quantity | 1 g |
| Synonym | Pyridine-2,3,4,6-d4, 5-(methyl-d3)- (9 CI), 3-Picoline-d7, 3-Methylpyridine-d7, 3-(Methyl-d3)pyridine-2,4,5,6-d4, 3-Methylpyridine-D7 |
| InChI Formula | InChI=1 S/C6H7N/c1-6-3-2-4-7-5-6/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1nc([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(trideuteriomethyl)pyridine |
| Molecular Weight (g/mol) | 100.1696 |
| Formula Weight | 100.1018 g/mol |
| Show More |