Learn More
3,4-Dichlorotoluene-2,5,6-d3, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 3,4-Dichlorotoluene-2,5,6-D3 |
| CAS | 1219795-29-5 |
| Molecular Formula | C7 D3 H3 Cl2 |
| Quantity | 1 g |
| Synonym | 1,2-Dichloro-4-methylbenzene-3,5,6-d3, 3,4-Dichlorotoluene-d3, 3,4-Dichlorotoluene-2,5,6-d3, 3,4-Dichlorotoluene-2,5,6-D3, 4,5-Dichlorotoluene-2,3,6-d3 |
| InChI Formula | InChI=1 S/C7H6Cl2/c1-5-2-3-6(8)7(9)4-5/h2-4 H,1H3/i2D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(Cl)c(Cl)c([2 H])c1C |
| IUPAC Name | 1,2-dichloro-3,4,6-trideuterio-5-methylbenzene |
| Molecular Weight (g/mol) | 164.047 |
| Formula Weight | 163.0035 g/mol |
| Show More |