Learn More
2-Nonanone-1,1,1,3,3-d5, CDN
Product Code. 30324496
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30324496
Brand: CDN CDND74391G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Nonanone-1,1,1,3,3-D5
CAS 1398065-76-3
Molecular Formula C9 D5 H13 O
Quantity 1 g
Synonym 2-Nonanone-d5, 2-Nonanone-1,1,1,3,3-D5, 2-Nonanone-1,1,1,3,3-d5, Heptyl-1,1-d2 methyl-d3 ketone
InChI Formula InChI=1 S/C9H18O/c1-3-4-5-6-7-8-9(2)10/h3-8H2,1-2H3/i2D3,8D2
SMILES [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])CCCCCC
IUPAC Name 1,1,1,3,3-pentadeuteriononan-2-one
Molecular Weight (g/mol) 147.2694
Formula Weight 147.1671 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity