missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylpropionic-d7 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Isobutyric acid-d7 |
| CAS | 223134-74-5 |
| Molecular Formula | C4 2H7 H O2 |
| Quantity | 1 g |
| Synonym | Propanoic-2,3,3,3-d4 acid, 2-(methyl-d3)-, 2-(Methyl-d3)propanoic-2,3,3,3-d4 acid, Isobutyric acid D7, 2-Methyl-1-propanoic Acid-d7, 2-Methylpropanoic Acid-d7, 2-Methylpropionic Acid-d7, 2-Propanecarboxylic Acid-d7, Dimethylacetic Acid-d7, Isobutanoic Acid-d7, Isopropylformic Acid-d7, NSC 62780-d7, i-Butyric Acid-d7, iso-Butyric Acid-d7, α-Methylpropanoic Acid-d7, α-Methylpropionic Acid-d7 |
| InChI Formula | InChI=1 S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)/i1D3,2D3,3 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C(=O)O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,3,3-tetradeuterio-2-(trideuteriomethyl)propanoic acid |
| Molecular Weight (g/mol) | 95.15 |
| Formula Weight | 95.0964 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction