Learn More
2-Hexanone-1,1,1,3,3-d5, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Hexanone-1,1,1,3,3-d5 |
| CAS | 4840-82-8 |
| Molecular Formula | C6 D5 H7 O |
| Quantity | 1 g |
| Synonym | 2-Hexanone-1,1,1,3,3-d5 (7 CI, 8 CI, 9 CI, ACI), 2-Hexanone-1,1,1,3,3-D5, 2-Hexanone-d5, 2-Oxohexane-1,1,1,3,3-d5, Butyl methyl ketone-d5, Methyl butyl ketone-d5 |
| InChI Formula | InChI=1 S/C6H12O/c1-3-4-5-6(2)7/h3-5H2,1-2H3/i2D3,5D2 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])CCC |
| IUPAC Name | 1,1,1,3,3-pentadeuteriohexan-2-one |
| Molecular Weight (g/mol) | 105.19 |
| Formula Weight | 105.12 g/mol |
| Show More |