missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Iodobutane-2,2,3,3,4,4,4-d7 (Stabilized with copper), CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Iodobutane-2,2,3,3,4,4,4-D7 |
| CAS | 2708278-33-3 |
| Molecular Formula | C4 D7 H2 I |
| Quantity | 1 g |
| Synonym | 1-Iodobutane-2,2,3,3,4,4,4-d7, 4-Iodobutane-1,1,1,2,2,3,3-d7, 1-Iodobutane-2,2,3,3,4,4,4-D7, Butyl-2,2,3,3,4,4,4-d7 iodide, Butyl iodide-d7 |
| InChI Formula | InChI=1 S/C4H9I/c1-2-3-4-5/h2-4H2,1H3/i1D3,2D2,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CI |
| IUPAC Name | 1,1,1,2,2,3,3-heptadeuterio-4-iodobutane |
| Molecular Weight (g/mol) | 191.0619 |
| Formula Weight | 191.0188 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction