missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopropane-d7, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromopropane-d7 |
| CAS | 61909-26-0 |
| Molecular Formula | C3 D7 Br |
| Quantity | 1 g |
| Synonym | 1-Propyl Bromide-d7, 3-Bromopropane-d7, Abzol JG-d7, Ascusol MC-d7, Drysolv-d7, Leksol-d7, Propyl Bromide-d7, n-Propyl Bromide-d7, 3-Bromopropane-1,1,1,2,2,3,3-d7, Heptadeuteriopropyl Bromide, Propyl-d7 Bromide |
| InChI Formula | InChI=1 S/C3H7Br/c1-2-3-4/h2-3H2,1H3/i1D3,2D2,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1-bromo-1,1,2,2,3,3,3-heptadeuteriopropane |
| Molecular Weight (g/mol) | 130.03 |
| Formula Weight | 129.0171 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction