Learn More
1,3-Dichloroacetone-d4, CDN
Product Code. 30328695
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30328695
Brand: CDN CDND50301G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,3-Dichloroacetone-d4
CAS 350818-52-9
Molecular Formula C3 2H4 Cl2 O
Quantity 1 g
Synonym 2-Propanone-1,1,3,3-d4, 1,3-dichloro- (9 CI), 1,3-Dichloroacetone-d4, 1,3-Dichloro-1,1,3,3-tetradeuteriopropan-2-one
InChI Formula InChI=1 S/C3H4Cl2O/c4-1-3(6)2-5/h1-2H2/i1D2,2D2
SMILES [2 H]C([2 H])(Cl)C(=O)C([2 H])([2 H])Cl
IUPAC Name 1,3-dichloro-1,1,3,3-tetradeuteriopropan-2-one
Molecular Weight (g/mol) 130.994
Formula Weight 129.989 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity