Learn More
1cP-LSD, TRC
Product Code. 30136683
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136683 10 mg 10mg
30005777 1 mg 1mg
30012489 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136683 Supplier Toronto Research Chemicals Supplier No. TRCL35502510MG
 Restricted Item
Estimated Shipment: 23-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula C24H29N3O2
Molecular Weight (g/mol) 391.51
Synonym (6aR,9R)-4-(Cyclopropanecarbonyl)-N,N-diethyl-7-methyl-4,6,6a,7,8,9-hexahydroindolo[4,3-fg]quinoline-9-carboxamide, 1cP-LSD
IUPAC Name (6aR,9R)-4-(cyclopropanecarbonyl)-N,N-diethyl-7-methyl-6,6a,8,9-tetrahydroindolo[4,3-fg]quinoline-9-carboxamide
SMILES CCN(CC)C(=O)[C@H]1CN(C)[C@@H]2Cc3cn(C(=O)C4CC4)c5cccc(C2=C1)c35
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C24H29N3O2/c1-4-26(5-2)23(28)17-11-19-18-7-6-8-20-22(18)16(12-21(19)25(3)13-17)14-27(20)24(29)15-9-10-15/h6-8,11,14-15,17,21H,4-5,9-10,12-13H2,1-3H3/t17-,21-/m1/s1
Formula Weight 391.226
Chemical Name or Material 1CP-LSD (1-Cyclopropionyl lysergic acid diethylamide)